Learn More
Sino Biological SARS-CoV-2 Spike RBD (L452R,T478K) Protein (His Tag)
Supplier:  Sino Biological 40592V08H90100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike RBD (YP_009724390.1, with mutations L452R,T478K) (Arg319-Phe541) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.617.2) which emerged in the India.
Catalog No. 50-211-4323