Learn More
Sino Biological SARS-CoV-2 Spike RBD (P479L) Protein (His Tag) 100ug
Catalog No. 502114307
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike RBD (YP_009724390.1, with mutation P479L) (Arg319-Phe541) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-4307 Supplier Sino Biological Supplier No. 40592V08H100100
A DNA sequence encoding the SARS-CoV-2 Spike RBD (YP_009724390.1, with mutation P479L) (Arg319-Phe541) was expressed with a polyhistidine tag at the C-terminus.