Learn More
Cayman Chemical SARSCoV2 SpIke Reagent 100dtn
SDP
Catalog No. 502893932
Encompass_Preferred
SARS-CoV-2 Spike Reagent has been tested and formulated to work exclusively with Cayman’s SARS-CoV-2 Neutralizing Antibody SimpleDetect ELISA Kit (Item No. 502220).
Catalog No. 50-289-3932 Supplier Cayman Chemical Supplier No. 502221100 DTN
SARS-CoV-2 Spike Reagent has been tested and formulated to work exclusively with Cayman’s SARS-CoV-2 Neutralizing Antibody SimpleDetect ELISA Kit (Item No. 502220).