Learn More
Cayman Chemical SARSCoV2 SpIke Reagent120uL
SDP
Catalog No. 502896100
Encompass_Preferred
A quenched fluorescent probe for GST activity can be used in adherent or suspension cells cell or tissue lysates or purified enzyme preparations upon deprotection by GSTs rhodamine 110 is released and its fluorescence can be quantified using ex/em maxima of 496/520 nm respectively
Catalog No. 50-289-6100 Supplier Cayman Chemical Supplier No. 502071120 UL
A quenched fluorescent probe for GST activity can be used in adherent or suspension cells cell or tissue lysates or purified enzyme preparations upon deprotection by GSTs rhodamine 110 is released and its fluorescence can be quantified using ex/em maxima of 496/520 nm respectively