Learn More
Sino Biological SARS-CoV-2 Spike S1 (E154K, L452R, E484Q, D614G, P681R) Protein (His Tag)
Supplier:  Sino Biological 40591V08H19100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1 (YP_009724390.1, with mutations E154K, L452R, E484Q, D614G, P681R) (Met1-Arg685) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.617) which emerged in the India.
Catalog No. 50-224-7024