Learn More
Sino Biological SARS-CoV-2 Spike S1 (G75V, T76I, R246N, 247 deletion, 253 deletion, L452Q, F490S, D614G) Protein (His Tag)
Supplier:  Sino Biological 40591V08H31100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1 (YP_009724390.1, with mutations G75V, T76I, R246N, 247 deletion, 253 deletion, L452Q, F490S, D614G) (Met1-Arg685) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant C.37) which emerged in the Peru.
Catalog No. 50-224-7059