Learn More
Sino Biological SARS-CoV-2 Spike S1 ((LAL242-244)deletion, L18F, D80A, D215G, R246I, K417N, E484K, N501Y, D614G) Protein (His & AVI Tag), Biotinylated
Supplier:  Sino Biological 40591V49H5B20
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1 (YP_009724390.1, with mutations (LAL242-244)deletion, L18F, D80A, D215G, R246I, K417N, E484K, N501Y, D614G) (Met1-Arg685) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The purified protein was biotinylated in vitro. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.351) which emerged in the South Africa.
Catalog No. 50-224-7080