Learn More
Sino Biological SARS-CoV-2 Spike S1 NTD (T19R, T95I, G142D, 156-157 deletion, R158G) Protein (His Tag)
Supplier:  Sino Biological 40591V08H36100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1 NTD (YP_009724390.1, with mutations T19R, T95I, G142D, 156-157 deletion, R158G) (Met1-Ser305) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.617.2) which emerged in the India.
Catalog No. 50-224-6836