Learn More
Sino Biological SARS-CoV-2 Spike S1 (Q52R, A67V, 69-70 deletion, 144 deletion, E484K, D614G, Q677H) Protein (His Tag)
Supplier:  Sino Biological 40591V08H29100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1 (YP_009724390.1, with mutations Q52R, A67V, 69-70 deletion, 144 deletion, E484K, D614G, Q677H) (Met1-Arg685) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.525) which emerged in the Multiple countries.
Catalog No. 50-224-6834