Learn More
Sino Biological SARS-CoV-2 Spike S1 (T19R, G142D, L452R, E484Q, D614G, P681R) Protein (His & AVI Tag), Biotinylated
Supplier:  Sino Biological 40591V49H3B20
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1 (YP_009724390.1, with mutations T19R, G142D, L452R, E484Q, D614G, P681R) (Met1-Arg685) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The purified protein was biotinylated in vitro. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.617.3) which emerged in the India.
Catalog No. 50-224-7128