Learn More
Sino Biological SARS-CoV-2 Spike S1 (T95I, G142D, E154K, L452R, E484Q, D614G, P681R) Protein (His & AVI Tag), Biotinylated
Supplier:  Sino Biological 40591V49H1B20
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1 (YP_009724390.1, with mutations T95I, G142D, E154K, L452R, E484Q, D614G, P681R) (Met1-Arg685) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed. The mutations were identified in the SARS-CoV-2 variant (known as B.1.617.1) which emerged in the India.
Catalog No. 50-224-6873