Learn More
Sino Biological SARS-CoV-2 Spike S1 (T95I, Y144S, Y145N, R346K, E484K, N501Y, D614G, P681H) Protein (His Tag)
Supplier:  Sino Biological 40591V08H38100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1 (YP_009724390.1, with mutations T95I, Y144S, Y145N, R346K, E484K, N501Y, D614G, P681H) (Met1-Arg685) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.621) which emerged in the South America.
Catalog No. 50-224-6837