Learn More
Sino Biological SARS-CoV-2 Spike S1+S2 (D80A, LAL242-244 deletion, R246I, K417N, E484K, N501Y, D614G, A701V) Protein (ECD, His Tag) 100ug
Supplier:  Sino Biological 40589V08B11100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1+S2 (YP_009724390.1, with mutations D80A, LAL242-244 deletion, R246I, K417N, E484K, N501Y, D614G, A701V) (Met1-Pro1213) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as 20C/501Y.V2 or B.1.351 lineage) which emerged in South Africa.
Catalog No. 50-211-4275