Learn More
Sino Biological SARS-CoV-2 Spike S1+S2 (E154K, L452R, E484Q, D614G, P681R, E1072K, K1073R) Protein (ECD, His Tag) 100ug
Supplier:  Sino Biological 40589V08B12100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1+S2 (YP_009724390.1, with mutations E154K, L452R, E484Q, D614G, P681R, E1072K, K1073R) (Met1-Pro1213) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as B.1.617) which emerged in the India
Catalog No. 50-211-4276