Learn More
Sino Biological SARS-CoV-2 Spike S1+S2 (G75V, T76I, RSYLTPD246-252 deletion, D253N, L452Q, F490S, D614G, T859N) Protein (ECD, His Tag)
Supplier:  Sino Biological 40589V08B24100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1+S2 (YP_009724390.1, with mutations G75V, T76I, RSYLTPD246-252 deletion, D253N, L452Q, F490S, D614G, T859N) (Met1-Pro1213) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant C.37) which emerged in the Peru.
Catalog No. 50-224-7071