Learn More
Sino Biological SARS-CoV-2 Spike S1+S2 (L5F, T95I, D253G, S477N, E484K, D614G, A701V) Protein (ECD, His Tag)
Supplier:  Sino Biological 40589V08B21100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1+S2 (YP_009724390.1, with mutations L5F, T95I, D253G, S477N, E484K, D614G, A701V) (Met1-Pro1213) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.526) which emerged in the United States of America.
Catalog No. 50-224-6881