Learn More
Sino Biological SARS-CoV-2 B.1.1.529 (Omicron) Nucleocapsid Protein (His Tag) 100ug
Supplier:  Sino Biological 40588V07E34100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Nucleocapsid (YP_009724397.2, with mutations P13L,¿ERS31-33 deletion,¿R203K,¿G204R) (Met1-Ala419) was expressed with a polyhistidine tag at the N-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.1.529) which emerged in the South Africa.
Catalog No. 50-222-1636