Learn More
Epigentek Inc Recombinant SARS-CoV-2 Spike S1 Protein with His and Avi Tag (100 µg)
SDP
Supplier:  Epigentek Inc E800031
Encompass_Preferred
Recombinant SARS-CoV-2 Spike S1 Protein is produced by HEK293 cells expression system. The target protein is expressed with sequence (Gln14-Arg683) of 2019-ncov Spike S1 (Accession #YP_009724390.1) fused with a 6xHis tag and Avi at the C-terminus.
Catalog No. 50-201-9350