Learn More
Cayman Chemical SARSCoV2 SurfAC Glycoprote
SDP
Catalog No. 502947476
Encompass_Preferred
Immunogen: Recombinant SARS-CoV-2 Surface Glycoprotein RBD-mFc Protein • Host: HEK293 cells • Species Reactivity: (+) SARS-CoV-2, HCoV-HKU1 (isolate N1); (-) SARS-CoV, MERS-CoV, HCoV-HKU1 (isolate N5), HCoV-NL63, HCoV-229E, HCoV-OC4 • Applications: ELISA
Catalog No. 50-294-7476 Supplier Cayman Chemical Supplier No. 33627100 UG
Immunogen: Recombinant SARS-CoV-2 Surface Glycoprotein RBD-mFc Protein • Host: HEK293 cells • Species Reactivity: (+) SARS-CoV-2, HCoV-HKU1 (isolate N1); (-) SARS-CoV, MERS-CoV, HCoV-HKU1 (isolate N5), HCoV-NL63, HCoV-229E, HCoV-OC4 • Applications: ELISA