Learn More
Sino Biological SARS-CoV-2 XD(BA.1 x AY.4) Spike S1+S2 trimer Protein (ECD, His Tag)
Supplier:  Sino Biological 40589V08H30100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 XD(BA.1 x AY.4) Spike S1+S2 trimer (YP_009724390.1, with mutations T19R, A27S, T95I, G142D, EFR156G, NL211I, Ins214EPE, G339D, S371L, S373P, S375F, K417N, N440K, G446S, S477N, T478K, E484A, Q493R, G496S, Q498R, N501Y, Y505H, T547K, D614G, H655Y, N679K, P681H, N764K, D796Y, F817P, N856K, A892P, A899P, A942P, Q954H, N969K, L981F, K986P, V987P and furin cleavage site mutants) expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant XD(BA.1 x AY.4)).
Catalog No. 50-224-6483