Learn More
Cayman Chemical SARSCoV2B 1 1 7Alpha Va 96
SDP
Supplier:  Cayman Chemical 50218096 SOLID
Encompass_Preferred
Cayman’s SARS-CoV-2 (B.1.1.7, Alpha Variant) Neutralizing Antibody SimpleDetect ELISA Kit is a competitive assay that can be used for qualitative and/or semi-quantitative measurement of neutralizing antibodies in human plasma and serum. The assay has a range of 7.81-1,000 ng/ml, with a midpoint (50% B/B0) of 100-160 ng/ml, and a sensitivity (80% B/B0) of approximately 49 ng/ml.
Catalog No. 50-289-3197