Learn More
Cayman Chemical SARSCoVSARSCoV2 Nucleocap
SDP
Supplier:  Cayman Chemical 30822100 UL
Encompass_Preferred
An a-AR antagonist inhibits norepinephrine-induced contractions in rat vas deferens guinea pig spleen and rat aorta regions that express high levels of a1A-AR a1B-AR and a1D-ARs respectively (pA2s 7 93 7 13 and 8 26 respectively) inhibits histamine- angiotensin II- pentagastrin- carbachol and serotonin-induced contractions in guinea pig ileum in a dose-dependent manner reverses pupil dilation induced by the combination of phenylephrine and tropicamide in rabbits
Catalog No. 50-298-9826