Learn More
Cayman Chemical SARSCoVSARSCoV2 Nucleocap
SDP
Catalog No. 502991890
Encompass_Preferred
An alkaloid active against the F32 chloroquine-sensitive (IC50 0 52 UM) and FcB1 and FcM2 chloroquine-resistant strains of P falciparum (IC50s 0 52 0 8 and 0 49 UM respectively) and reduces parasitemia in a mouse model of P vinckei infection (ED50 18 9 mg/kg per day) decreases LPS-induced increases in TNF-a IL-1B and IL-6 levels and p65 nuclear translocation in RAW 264 7 cells at 5 UM decreases serum levels of IL-10 TNF-a and IL-6 and reduces mortality in a mouse model of LPS-induced endotoxemia inhibits proliferation of various cancer cells (IC50s 2 65-370 ng/ml) induces apoptosis and decreases STAT3 phosphorylation in HSC3 and HSC4 oral cancer cells from 0 1-10 UM and reduces tumor growth in an HSC3 mouse xenograft model at 10 mg/kg per day for 24 days
Catalog No. 50-299-1890 Supplier Cayman Chemical Supplier No. 31428100 UL
An alkaloid active against the F32 chloroquine-sensitive (IC50 0 52 UM) and FcB1 and FcM2 chloroquine-resistant strains of P falciparum (IC50s 0 52 0 8 and 0 49 UM respectively) and reduces parasitemia in a mouse model of P vinckei infection (ED50 18 9 mg/kg per day) decreases LPS-induced increases in TNF-a IL-1B and IL-6 levels and p65 nuclear translocation in RAW 264 7 cells at 5 UM decreases serum levels of IL-10 TNF-a and IL-6 and reduces mortality in a mouse model of LPS-induced endotoxemia inhibits proliferation of various cancer cells (IC50s 2 65-370 ng/ml) induces apoptosis and decreases STAT3 phosphorylation in HSC3 and HSC4 oral cancer cells from 0 1-10 UM and reduces tumor growth in an HSC3 mouse xenograft model at 10 mg/kg per day for 24 days