Learn More
Cayman Chemical SARSCoVSARSCoV2 Nucleocap 50uL
SDP
Catalog No. 502988882
Encompass_Preferred
An inhibitor of SMYD2 (IC50 1 7 M) selective for SMYD2 over PAR1 (IC50 30 M)
Catalog No. 50-298-8882 Supplier Cayman Chemical Supplier No. 3082250 UL
An inhibitor of SMYD2 (IC50 1 7 M) selective for SMYD2 over PAR1 (IC50 30 M)