Learn More
Cayman Chemical SARSCoVSARSCoV2 Nucleocap 50uL
SDP
Supplier:  Cayman Chemical 3198750 UL
Encompass_Preferred
12(S)-hydroxy-16-Heptadecynoic acid is a mechanism-based inhibitor of CYP450 W-hydroxylase.{1051} It inhibits prostaglandin W-hydroxylase with a Ki value of 1.8 UM.{1051}
Catalog No. 50-294-7510