Learn More
Cayman Chemical SARSCoVSARSCoV2 SpIke Gly
SDP
Catalog No. 502947786
Encompass_Preferred
An internal standard for the quantification of rilmenidine by GC- or LC-MS
Catalog No. 50-294-7786 Supplier Cayman Chemical Supplier No. 31988100 UL
An internal standard for the quantification of rilmenidine by GC- or LC-MS