Learn More
Cayman Chemical SARSCoVSARSCoV2 SpIke Gly
SDP
Supplier:  Cayman Chemical 31989100 UL
Encompass_Preferred
An internal standard for the quantification of rilmenidine by GC- or LC-MS
Catalog No. 50-294-8754