Learn More
Sartorius Balance Dot-Matrix Printers
Catalog No. 01913377
Change view
Click to view available options
Printer D1 Series 115v 60hz
Catalog No.
01-913-377 Printer D1 Series 115v 60hz
1 options
Catalog No. 01-913-377 Supplier Sartorius Supplier No. 901041.1
Includes
Supplied AC adapter/charger also permits AC operation at 115V 50/60Hz. Comes with connection cable, one roll of paper, and one purple ink cartridge.
Dot-Matrix Printer. For Sartorius balances. Ink ribbon. Prints up to 1 3/4 lines per second; 40 characters per line. Standard RS-232-C interface. Adjustable baud rate between 300 and 19,200bps. Features full dot addressable graphics, alphanumeric characters (including an international character set) for printing weighing units or statistical data, and an integral self-test and diagnostic mode.
Order Info
Unit measures 7 3/4L x 4 1/8W x 3 1/2 in.H (19.7 x 10.5 x 8.9cm). Net weight 1 3/8 lb. (0.6kg).
Specifications
For Use With (Equipment) DI, TL, TR, TC Series
Type Dot Matrix Printer