Learn More
Chemglass Life Sciences Sartorius Motor Adapter Kit
SDP
Catalog No. 502733318
Encompass
Adapter kit to convertstandard Applikon style agitator assembly to fit a Satorius motor.NOTE: This is the ADAPTER KIT ONLY, it does not include the agitator assembly.
Catalog No. 50-273-3318 Supplier Chemglass Life Sciences Supplier No. CLS1380SAR1
Adapter kit to convertstandard Applikon style agitator assembly to fit a Satorius motor.NOTE: This is the ADAPTER KIT ONLY, it does not include the agitator assembly.