Learn More
Cayman Chemical Sartrypyron B 10mg
SDP
Catalog No. 502992011
Encompass_Preferred
A fungal metabolite; inhibits the growth of MCF-7, H460, and A375 cancer cells (GI50s = 17.8, 20.5, and 25 µM, respectively)
Catalog No. 50-299-2011 Supplier Cayman Chemical Supplier No. 2994910 MG
A fungal metabolite; inhibits the growth of MCF-7, H460, and A375 cancer cells (GI50s = 17.8, 20.5, and 25 µM, respectively)