Learn More
Selleck Chemical LLC Saruparib S9875-25MG
SDP
Catalog No. 5000024677
Encompass_Preferred
Saruparib (AZD5305) is a highly selective and potent inhibitor of PARP1 with an IC50 of 3 nM in wild-type A549 lung cancer cells AZD5305 shows no or minimal growth inhibitory effects in other cells (IC50s 10 M)
Catalog No. 50-000-24677 Supplier Selleck Chemical LLC Supplier No. S987525MG
Saruparib (AZD5305) is a highly selective and potent inhibitor of PARP1 with an IC50 of 3 nM in wild-type A549 lung cancer cells AZD5305 shows no or minimal growth inhibitory effects in other cells (IC50s 10 M)