Learn More
Selleck Chemical LLC Saruparib AZD5305-5mg
SDP
Supplier:  Selleck Chemical LLC S98755MG
Encompass_Preferred
Saruparib (AZD5305) is a highly selective and potent inhibitor of PARP1 with an IC50 of 3 nM in wild-type A549 lung cancer cells. AZD5305 shows no or minimal growth inhibitory effects in other cells (IC50s >10μM).
Catalog No. 50-238-9585