Learn More
Bioscience International Inc SAS 7 Cell NiMH Rechargeable Battery, 2700 mAh Capacity, 8.4 V Voltage, Unit: Each
SDP
Catalog No. NC2351224
Encompass
The SAS 7 Cell NiMH Rechargeable Battery from BIOSCIENCE INTERNATIONAL INC is expertly designed for scientific applications, particularly to power SAS series microbial air samplers. It ensures reliable and extended performance with an 8.4 V voltage and a nominal capacity of 2700 mAh, supporting 7 hours of continuous operation or sampling of 40,000 liters. This functionality is crucial for environmental monitoring across pharmaceutical, food, and laboratory sectors, distinguishing it from standard batteries.
Catalog No. NC2351224 Supplier Bioscience International Inc Supplier No. 18200
The SAS 7 Cell NiMH Rechargeable Battery from BIOSCIENCE INTERNATIONAL INC is expertly designed for scientific applications, particularly to power SAS series microbial air samplers. It ensures reliable and extended performance with an 8.4 V voltage and a nominal capacity of 2700 mAh, supporting 7 hours of continuous operation or sampling of 40,000 liters. This functionality is crucial for environmental monitoring across pharmaceutical, food, and laboratory sectors, distinguishing it from standard batteries.