Learn More
Bioscience International Inc SAS Disposable Sampling Head, Sterile, Compatible with 90 mm Petri Dishes, Not Autoclaving Required
SDP
Catalog No. NC0554046
Encompass
The SAS Disposable Sampling Head, Petri Style, by Bioscience International Inc. offers a sterile, single-use solution for microbial air sampling. Eliminating autoclaving needs enhances lab workflow, while its compatibility with standard 90 mm Petri dishes ensures versatility. Designed for 100% sampling efficiency of airborne particles as small as 1 micron, it's perfect for controlled environments and cleanrooms. Each unit is individually packaged for sterility, making it a reliable choice for precise environmental monitoring.
Catalog No. NC0554046 Supplier Bioscience International Inc Supplier No. 91214
The SAS Disposable Sampling Head, Petri Style, by Bioscience International Inc. offers a sterile, single-use solution for microbial air sampling. Eliminating autoclaving needs enhances lab workflow, while its compatibility with standard 90 mm Petri dishes ensures versatility. Designed for 100% sampling efficiency of airborne particles as small as 1 micron, it's perfect for controlled environments and cleanrooms. Each unit is individually packaged for sterility, making it a reliable choice for precise environmental monitoring.