Learn More
Bioscience International Inc SAS Isolator Microbial Air Sampler, High-Volume 180 L/min, Programmable Base Unit, Portable
SDP
Catalog No. NC3460886
Encompass
The SAS Isolator is a high-volume microbial air sampler designed for critical monitoring in isolator and barrier environments. Its advanced impaction sampling method captures microorganisms efficiently, ensuring compliance with international standards. With a primary air flow rate of 180 L/min and a fast sampling time of under 6 minutes for 1,000 L of air, it offers reliability and speed. Its portable design includes long-lasting rechargeable batteries, making it ideal for cleanrooms and hospitals.
Catalog No. NC3460886 Supplier Bioscience International Inc Supplier No. 43217
Add to Cart
Add to Cart
Request a Quote
The SAS Isolator is a high-volume microbial air sampler designed for critical monitoring in isolator and barrier environments. Its advanced impaction sampling method captures microorganisms efficiently, ensuring compliance with international standards. With a primary air flow rate of 180 L/min and a fast sampling time of under 6 minutes for 1,000 L of air, it offers reliability and speed. Its portable design includes long-lasting rechargeable batteries, making it ideal for cleanrooms and hospitals.