Learn More
Bioscience International Inc SAS Super 100 Microbial Air Sampler, 100 L/min Airflow, 7 Hours Battery Life
SDP
Catalog No. NC0776812
Encompass
The SAS Super 100 is a portable microbial air sampler ideal for detecting airborne microbial contamination in critical environments like cleanrooms and hospitals. With a high airflow rate of 100 L/min and rapid sampling capability, it offers efficient sample collection on standard media. It features a robust electronic data management system and meets various international standards, ensuring reliability and traceability. Its long battery life of 7 hours distinguishes it from competitors, making it perfect for extensive testing scenarios.
Catalog No. NC0776812 Supplier Bioscience International Inc Supplier No. 18198
Add to Cart
Add to Cart
Request a Quote
The SAS Super 100 is a portable microbial air sampler ideal for detecting airborne microbial contamination in critical environments like cleanrooms and hospitals. With a high airflow rate of 100 L/min and rapid sampling capability, it offers efficient sample collection on standard media. It features a robust electronic data management system and meets various international standards, ensuring reliability and traceability. Its long battery life of 7 hours distinguishes it from competitors, making it perfect for extensive testing scenarios.