Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SASPA (Saturated Aqueous Picric Acid with Surfactant), Reagents

Catalog No. 01333516
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-516 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-516 Supplier Reagents Holdings Llc Supplier No. CS250600250C

missing translation for 'validMainframeMsg'

SASPA (Saturated Aqueous Picric Acid with Surfactant), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 88-89-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Saturated picric Acid
IUPAC Name water
SMILES O

Specifications

Color Yellow
Physical Form Liquid
Boiling Point 100°C
Quantity 250 mL
Packaging Glass Bottle
Specific Gravity 1.01
Grade Reagent
Chemical Name or Material SASPA (Saturated Aqueous Picric Acid with Surfactant)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.