Learn More
Aviva Systems Biology Corp SASS6 Antibody - C-terminal region (ARP62500_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62500_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SASS6. It was validated on Western Blot. . Alternate names: FLJ22097, MGC119440, SAS-6, SAS6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-697