Learn More
Boston Bioproducts Inc Saturated Ammonium Sulfate (~4.1 M) - 100ml
Catalog No. NC2384285
Encompass
The 4.1M Ammonium Sulfate Solution is widely utilized in biochemistry and molecular biology for precipitation and fractionation of proteins, hydrophobic interaction chromatography, and purification of antibodies. This solution is also commonly used in applications such as long PCR, second-strand cDNA synthesis, and HPLC analysis of phospholipids. The use of high concentrations of ammonium sulfate has been shown to inhibit microbial growth and maintain the protein in a folded state.
Catalog No. NC2384285 Supplier Boston Bioproducts Inc Supplier No. MT127100ML
The 4.1M Ammonium Sulfate Solution is widely utilized in biochemistry and molecular biology for precipitation and fractionation of proteins, hydrophobic interaction chromatography, and purification of antibodies. This solution is also commonly used in applications such as long PCR, second-strand cDNA synthesis, and HPLC analysis of phospholipids. The use of high concentrations of ammonium sulfate has been shown to inhibit microbial growth and maintain the protein in a folded state.