Learn More
Abcam SATA (N-Succinimidyl-S-acetylthioacetate), Protein modification thiolating reagent, 100MG
Catalog No. 502705103
Encompass_Preferred
MW 231.23 Da, Purity >94%. Primary amine-reactive protein modification thiolating reagent. Adds protected sulfhydryls to targeted proteins. Induces stable recombinant TNF-α trimer formation by formation of disulfide bridges between reactive thiol groups on adjacent TNF-α molecules, which show increased therapeutic efficacy in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5103 Supplier Abcam Supplier No. AB145622100MG
MW 231.23 Da, Purity >94%. Primary amine-reactive protein modification thiolating reagent. Adds protected sulfhydryls to targeted proteins. Induces stable recombinant TNF-α trimer formation by formation of disulfide bridges between reactive thiol groups on adjacent TNF-α molecules, which show increased therapeutic efficacy in vivo. .
The product is subject to the following: Abcam Restricted Use Statement