Learn More
Aviva Systems Biology Corp SATB1 Antibody - C-terminal region (ARP38208_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38208_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SATB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SATB1. Alternate names: ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56564