Learn More
Cayman Chemical SATB2CTerm RabbIt Monclo 100uL
SDP
Catalog No. 502947379
Encompass_Preferred
An inhibitor of B-RAF and C-RAF (IC50s 41 7 and 2 nM for wild-type B-RAF B-RAFV600E and wild-type C-RAF respectively) cytotoxic to B-RAFV600E-expressing A375 and SK-MEL-28 melanoma cells (IC50s 57 and 69 nM respectively) as well as COLO 205 and HCT116 colon and B-CPAP and CAL-62 thyroid cancer cells (IC50s 116 65 43 and 479 nM respectively) reduces tumor growth in A375 SK-MEL-28 SK-MEL-2 and SK-MEL-30 mouse xenograft models
Catalog No. 50-294-7379 Supplier Cayman Chemical Supplier No. 32290100 UL
An inhibitor of B-RAF and C-RAF (IC50s 41 7 and 2 nM for wild-type B-RAF B-RAFV600E and wild-type C-RAF respectively) cytotoxic to B-RAFV600E-expressing A375 and SK-MEL-28 melanoma cells (IC50s 57 and 69 nM respectively) as well as COLO 205 and HCT116 colon and B-CPAP and CAL-62 thyroid cancer cells (IC50s 116 65 43 and 479 nM respectively) reduces tumor growth in A375 SK-MEL-28 SK-MEL-2 and SK-MEL-30 mouse xenograft models