Learn More
Enterprise Technology Solutions Satco Products, Inc. Bulb,LED,A19,10W,2700K,WarmWhite,4/PK
SDP
Catalog No. 5000829224
Encompass_Preferred
LED A19 bulb with a frosted finish features an energy-efficient design that uses nearly 80 percent less electricity than an incandescent bulb to minimize your operating costs. Nondimmable bulb with 800 lumens is approved for use in enclosed fixtures and provides up to 15,000 hours of warm white light. 120-volt, 10-watt bulb also features a 2,700 Kelvin temperature, 93 CRI and medium base.
Catalog No. 50-008-29224 Supplier Enterprise Technology Solutions Supplier No. SDNS28560
LED A19 bulb with a frosted finish features an energy-efficient design that uses nearly 80 percent less electricity than an incandescent bulb to minimize your operating costs. Nondimmable bulb with 800 lumens is approved for use in enclosed fixtures and provides up to 15,000 hours of warm white light. 120-volt, 10-watt bulb also features a 2,700 Kelvin temperature, 93 CRI and medium base.