Learn More
Enterprise Technology Solutions Satco Products, Inc. Bulb,LED,BR40,13W,2700K,WarmWhite
SDP
Catalog No. 5000829238
Encompass_Preferred
LED BR40 bulb with a frosted finish features an energy-efficient design that uses nearly 80 percent less electricity than an incandescent bulb to minimize your operating costs. LED bulb offers a 103-degree beam spread to illuminate your space with a warm white light. Dimmable bulb with 1075 lumens provides up to 25,000 hours of light. 120-volt, 13-watt bulb also features a 2,700 Kelvin temperature, 95 CRI and medium base.
Catalog No. 50-008-29238 Supplier Enterprise Technology Solutions Supplier No. SDNS29615
LED BR40 bulb with a frosted finish features an energy-efficient design that uses nearly 80 percent less electricity than an incandescent bulb to minimize your operating costs. LED bulb offers a 103-degree beam spread to illuminate your space with a warm white light. Dimmable bulb with 1075 lumens provides up to 25,000 hours of light. 120-volt, 13-watt bulb also features a 2,700 Kelvin temperature, 95 CRI and medium base.