Learn More
Enterprise Technology Solutions Satco Products, Inc. Lightbulb,A19,Energy-Saving,LED,NonDimmable,25000Hours,2700K,3W/9W/12W,120-Volt,280L/950L/1200L,WE
SDP
Catalog No. 5000829095
Encompass_Preferred
A19 bulb with a frosted finish is wattage-selectable (3W/9W/12W) and produces a warm, white light. Bulb features energy-saving LED technology, still producing 280L/950L/1200L, respectively. Long-life design provides up to 25,000 hours of use and delivers a 220-degree beam spread. This modern and sophisticated bulb elegantly illuminates any room in the house. Non-dimmable, 120-volt bulb features a 2700K Kelvin temperature and 80+ CRI. Medium base fits most lamps and fixtures.
Catalog No. 50-008-29095 Supplier Enterprise Technology Solutions Supplier No. SDNS9316
A19 bulb with a frosted finish is wattage-selectable (3W/9W/12W) and produces a warm, white light. Bulb features energy-saving LED technology, still producing 280L/950L/1200L, respectively. Long-life design provides up to 25,000 hours of use and delivers a 220-degree beam spread. This modern and sophisticated bulb elegantly illuminates any room in the house. Non-dimmable, 120-volt bulb features a 2700K Kelvin temperature and 80+ CRI. Medium base fits most lamps and fixtures.