Learn More
Enterprise Technology Solutions Satco Products, Inc. Tube,Fluorescent,T5,54W,3500K,NeutralWhite,40/CT
SDP
Catalog No. 5000789972
Encompass_Preferred
High-performance, 54-watt fluorescent tube delivers neutral white light and a high output of 5000 lumens for approximately 24,000 hours. Rated at 3500 Kelvin, the T5-shape tube light radiates neutral light and offers a CRI of 85 and a miniature bi-pin base. Actual length is 48-4/5", but it is considered 48". Fluorescent tube is TCLP compliant and meets RoHS standards.
Catalog No. 50-007-89972 Supplier Enterprise Technology Solutions Supplier No. SDNS8144
High-performance, 54-watt fluorescent tube delivers neutral white light and a high output of 5000 lumens for approximately 24,000 hours. Rated at 3500 Kelvin, the T5-shape tube light radiates neutral light and offers a CRI of 85 and a miniature bi-pin base. Actual length is 48-4/5", but it is considered 48". Fluorescent tube is TCLP compliant and meets RoHS standards.