Learn More
Enterprise Technology Solutions Satco Products, Inc. Tube,LED,G13,11W,4000K,1800Lumens,CoolWhite,4',25/CT
SDP
Catalog No. 5000828734
Encompass_Preferred
T8 LED Tube Bulb illuminates rooms with 1800 lumens of cool white light (4000 Kelvin) using just 11 watts of energy. Highly efficient, LED bulb delivers long-lasting performance for up to 50,000 hours. Dimmable bulb also features a medium bi-pin base (G13 ANSI Base) and 82 CRI.
Catalog No. 50-008-28734 Supplier Enterprise Technology Solutions Supplier No. SDNS49936R1
T8 LED Tube Bulb illuminates rooms with 1800 lumens of cool white light (4000 Kelvin) using just 11 watts of energy. Highly efficient, LED bulb delivers long-lasting performance for up to 50,000 hours. Dimmable bulb also features a medium bi-pin base (G13 ANSI Base) and 82 CRI.