Learn More
World Precision Instrument Satellite Air-Therm Heater for use with ECU-AIRTHERM-SAT-1W
SDP
Catalog No. 502538934
Encompass
The AirTherm SAT is an auxiliary heater designed to be used in Live Cell Imaging applications with microscopes that are fitted with a full microscope environmental chamber enclosure. It works in conjunction with an ECU (environmental control unit), which uses a PID control algorithm to maintain tightly controlled loops of heat for environment control.
Catalog No. 50-253-8934 Supplier World Precision Instrument Supplier No. AIRTHERMSAT1W
The AirTherm SAT is an auxiliary heater designed to be used in Live Cell Imaging applications with microscopes that are fitted with a full microscope environmental chamber enclosure. It works in conjunction with an ECU (environmental control unit), which uses a PID control algorithm to maintain tightly controlled loops of heat for environment control.