Learn More
World Precision Instrument Satellite Air-Therm Heater for use with ECU-AIRTHERM-SAT-2W
SDP
Catalog No. 502538933
Encompass
The AirTherm SAT is an auxiliary heater designed to be used in Live Cell Imaging applications with microscopes that are fitted with a full microscope environmental chamber enclosure. It works in conjunction with an ECU (environmental control unit), which uses a PID control algorithm to maintain tightly controlled loops of heat for environment control.
Catalog No. 50-253-8933 Supplier World Precision Instrument Supplier No. AIRTHERMSAT2W
Add to Cart
Add to Cart
Request a Quote
The AirTherm SAT is an auxiliary heater designed to be used in Live Cell Imaging applications with microscopes that are fitted with a full microscope environmental chamber enclosure. It works in conjunction with an ECU (environmental control unit), which uses a PID control algorithm to maintain tightly controlled loops of heat for environment control.