Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Satigrel 25mg | 111753-73-2 | 337.37 g/mol | C20H19NO4 | 25 MG
SDP

Catalog No. 5000229957
Encompass_Preferred

Satigrel is a research-grade small molecule inhibitor of prostaglandin-endoperoxide synthase 1 (PGHS1/COX-1) that reduces thromboxane A2 synthesis and inhibits platelet aggregation. Intended for preclinical biochemical and pharmacological research only; not for human or veterinary use.

  • Selective PGHS1 (COX-1) inhibitor (IC50: 0.081 μM).
  • Also shows activity against PGHS2 (IC50: 5.9 μM) and some phosphodiesterase isoforms.
  • Molecular formula C20H19NO4; molecular weight ≈337.37 g/mol.
  • High purity (99.85%) suitable for biochemical assays.
  • Available in small milligram pack sizes for laboratory research.
  • CAS number 111753-73-2 for unambiguous identification.

Catalog No. 50-002-29957 Supplier Medchemexpress LLC Supplier No. HY11456425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Satigrel is a research-grade small molecule inhibitor of prostaglandin-endoperoxide synthase 1 (PGHS1/COX-1) that reduces thromboxane A2 synthesis and inhibits platelet aggregation. Intended for preclinical biochemical and pharmacological research only; not for human or veterinary use.

  • Selective PGHS1 (COX-1) inhibitor (IC50: 0.081 μM).
  • Also shows activity against PGHS2 (IC50: 5.9 μM) and some phosphodiesterase isoforms.
  • Molecular formula C20H19NO4; molecular weight ≈337.37 g/mol.
  • High purity (99.85%) suitable for biochemical assays.
  • Available in small milligram pack sizes for laboratory research.
  • CAS number 111753-73-2 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.